1,3,5-Tris(bromomethyl)benzene 98%

Supplier: Ambeed
Danger

A412770-25G A412770-5G
77551-236EA 98.88 USD
77551-236 77568-832
1,3,5-Tris(bromomethyl)benzene 98%
1,3,5-Tris(bromomethyl)benzene

Precaución: For research use only.

Formula: C₉H₉Br₃
MW: 356.88 g/mol
MDL Number: MFCD00182500
CAS Number: 18226-42-1
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR