2,2-Dimethyl-4-oxo-3,8,11,14,17-pentaoxa-5-azanonadecan-19-oic acid 98%

Supplier: Ambeed
Warning

Synonyms: Boc-NH-PEG4-CH2COOH

A411835-5G A411835-1G A411835-100MG A411835-250MG A411835-25G
77551-232EA 300 USD
77551-232 77568-826 77594-262 77594-264 77748-831
2,2-Dimethyl-4-oxo-3,8,11,14,17-pentaoxa-5-azanonadecan-19-oic acid 98%
2,2-Dimethyl-4-oxo-3,8,11,14,17-pentaoxa-5-azanonadecan-19-oic acid

Precaución: For research use only.

Formula: C₁₅H₂₉NO₈
MDL Number: MFCD30528568
CAS Number: 876345-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR