8-Hydroxy-1,6-naphthyridine-7-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A403696-100MG A403696-1G A403696-250MG
77551-210EA 167.89 USD
77551-210 77748-815 77748-816
8-Hydroxy-1,6-naphthyridine-7-carboxylic acid ≥98%
8-Hydroxy-1,6-naphthyridine-7-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆N₂O₃
MW: 190.1555 g/mol
MDL Number: MFCD13151936
CAS Number: 410542-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR