Methyl-α-bromoisobutyrate ≥95%

Supplier: Ambeed
Danger

Synonyms: 2-Bromo-2-methylpropionic acid methyl ester, Methyl 2-Bromo-2-methylpropanoate, Methyl 2-bromo-2-methylpropionate, alpha-Bromoisobutyricacidmethylester, Methyl 2-bromo-2-methylpropanoate, Methyl α-bromoisobutyrate, Methyl 2-bromoisobutyrate

A399099-25G A399099-100G
77551-198EA 19.57 USD
77551-198 77594-216
Methyl-α-bromoisobutyrate ≥95%
Methyl-α-bromoisobutyrate

Precaución: for Research Use Only

Formula: C₅H₉BrO₂
MW: 181.029 g/mol
MDL Number: MFCD00017854
CAS Number: 23426-63-3
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR