N-Methyl-tert-butylamine ≥95%

Supplier: Ambeed
Danger

A392346-5G A392346-25G
77551-162EA 57.68 USD
77551-162 77572-188
N-Methyl-tert-butylamine ≥95%
N-Methyl-tert-butylamine

Precaución: for Research Use Only

Formula: C₅H₁₃N
MW: 87.166 g/mol
MDL Number: MFCD00042853
CAS Number: 14610-37-8
UN: 2733
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR