2,2,2-Trifluoro-N-phenylacetimidoyl chloride ≥98%

Supplier: Ambeed
Danger

A389164-5G A389164-10G A389164-25G
77551-152EA 88.58 USD
77551-152 77572-174 77572-176
2,2,2-Trifluoro-N-phenylacetimidoyl chloride ≥98%
2,2,2-Trifluoro-N-phenylacetimidoyl chloride

Precaución: for Research Use Only

Formula: C₈H₅ClF₃N
MW: 207.58 g/mol
MDL Number: MFCD11042687
CAS Number: 61881-19-4
UN: 2924
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR