Ethyl but-3-ynoate ≥95%
Supplier: Ambeed
|
Danger
|
Synonyms:
Ethyl3-Butynoate
A386202-1G
A386202-5G
77551-140EA
113.3
USD
77551-140
77668-794
Ethyl but-3-ynoate ≥95%
Ethyl but-3-ynoate
Precaución: for Research Use Only
Formula:
C₆H₈O₂ MW: 112.126 g/mol |
MDL Number:
MFCD07780250 CAS Number: 53841-07-9 UN: 1993 ADR: 3,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...