6-Fluoroquinoline-2-carboxylic acid ≥96%

Supplier: Ambeed
Warning

A386066-1G A386066-100MG
77551-138EA 131.84 USD
77551-138 77672-912
6-Fluoroquinoline-2-carboxylic acid ≥96%
6-Fluoroquinoline-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆FNO₂
MW: 191.159 g/mol
MDL Number: MFCD09837268
CAS Number: 86324-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR