5,5'-(1,4-Phenylenebis(oxy))bis(isobenzofuran-1,3-dione) ≥98%

Supplier: Ambeed
Warning

A382752-1G A382752-5G
77551-112EA 80.34 USD
77551-112 77594-146
5,5'-(1,4-Phenylenebis(oxy))bis(isobenzofuran-1,3-dione) ≥98%
5,5'-(1,4-Phenylenebis(oxy))bis(isobenzofuran-1,3-dione)

Precaución: for Research Use Only

Formula: C₂₂H₁₀O₈
MW: 402.31 g/mol
MDL Number: MFCD00346722
CAS Number: 17828-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR