4-Bromo-2-nitro-1H-imidazole ≥98%

Supplier: Ambeed
Warning

A380558-5G A380558-100MG A380558-250MG
77551-108EA 238.96 USD
77551-108 77672-888 77672-890
4-Bromo-2-nitro-1H-imidazole ≥98%
4-Bromo-2-nitro-1H-imidazole

Precaución: for Research Use Only

Formula: C₃H₂BrN₃O₂
MW: 191.97 g/mol
MDL Number: MFCD09028957
CAS Number: 121816-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR