Potassium thioacetate ≥97%

Supplier: Ambeed
Danger

Synonyms: Potassium ethanethioate

A379560-25G A379560-500G A379560-1000G A379560-100G
77551-104EA 13.39 USD
77551-104 77551-106 77572-128 77668-758
Potassium thioacetate ≥97%
Potassium thioacetate

Precaución: for Research Use Only

Formula: C₂H₃KOS
MW: 114.21 g/mol
MDL Number: MFCD00137704
CAS Number: 10387-40-3
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR