trans-Stilbene ≥98%

Supplier: Ambeed
Danger

Synonyms: trans-1,2-Diphenylethylene, (E)-1,2-Diphenylethene, trans-1,2-Diphenylethene

A373852-100G A373852-25G A373852-5G
77551-078EA 101 USD
77551-078 77551-080 77551-082
trans-Stilbene ≥98%
trans-Stilbene

Precaución: for Research Use Only

Formula: C₁₄H₁₂
MDL Number: MFCD00064300
CAS Number: 103-30-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR