N,N-Diethylcarbamoyl chloride ≥98%

Supplier: Ambeed
Danger

Synonyms: Diethylcarbamyl chloride

A372452-25G A372452-100G
77551-076EA 10 USD
77551-076 77626-720
N,N-Diethylcarbamoyl chloride ≥98%
N,N-Diethylcarbamoyl chloride

Precaución: for Research Use Only

Formula: C₅H₁₀ClNO
MW: 135.59 g/mol
MDL Number: MFCD00000636
CAS Number: 88-10-8
UN: 3265
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR