6-Amino-1H-indazole-7-carboxylic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: 6-Amino-7-carboxy-1H-indazole

A368781-1G A368781-100MG
77551-068EA 237.93 USD
77551-068 77672-838
6-Amino-1H-indazole-7-carboxylic acid ≥98%
6-Amino-1H-indazole-7-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MW: 177.16 g/mol
MDL Number: MFCD03856017
CAS Number: 73907-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR