5-Bromo-3-furoic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromofuran-3-carboxylic acid, 2-Bromo-4-carboxyfuran

A367647-5G A367647-250MG
77551-064EA 210.12 USD
77551-064 77746-736
5-Bromo-3-furoic acid ≥95%
5-Bromo-3-furoic acid

Precaución: for Research Use Only

Formula: C₅H₃BrO₃
MW: 190.98 g/mol
MDL Number: MFCD08669547
CAS Number: 58832-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR