Methyl bromodifluoroacetate ≥97%

Supplier: Ambeed
Danger

Synonyms: Bromodifluoroacetic Acid Methyl Ester

A359280-25G A359280-5G
77551-018EA 37.08 USD
77551-018 77551-020
Methyl bromodifluoroacetate ≥97%
Methyl bromodifluoroacetate

Precaución: for Research Use Only

Formula: C₃H₃BrF₂O₂
MW: 188.96 g/mol
MDL Number: MFCD06248118
CAS Number: 683-98-7
UN: 2924
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR