Trichloroacetic anhydride ≥98%

Supplier: Ambeed
Danger

A359054-25G A359054-5G
77551-016EA 37 USD
77551-016 77872-544
Trichloroacetic anhydride ≥98%
Trichloroacetic anhydride

Precaución: for Research Use Only

Formula: C₄Cl₆O₃
MDL Number: MFCD00000793
CAS Number: 4124-31-6
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR