(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-methylnon-8-enoic acid 95%

Supplier: Ambeed
Warning

A352583-1G A352583-5G
77551-004EA 517.06 USD
77551-004 77568-712
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-methylnon-8-enoic acid 95%
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-methylnon-8-enoic acid

Precaución: For research use only.

Formula: C₂₅H₂₉NO₄
MW: 407.502 g/mol
MDL Number: MFCD19381836
CAS Number: 1311933-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR