Adamantane-1,3,5,7-tetracarboxylic acid ≥95%

Supplier: Ambeed
Warning

A348967-1G A348967-100MG A348967-250MG
77551-002EA 366 USD
77551-002 77900-642 77901-040
Adamantane-1,3,5,7-tetracarboxylic acid ≥95%
Adamantane-1,3,5,7-tetracarboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₆O₈
MDL Number: MFCD09907630
CAS Number: 100884-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR