3-Methoxy-2,6-dimethylaniline ≥98%

Supplier: Ambeed
Warning

A344413-5G A344413-1G A344413-250MG
77550-994EA 333 USD
77550-994 77594-022 77746-674
3-Methoxy-2,6-dimethylaniline ≥98%
3-Methoxy-2,6-dimethylaniline

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MDL Number: MFCD08693118
CAS Number: 95645-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR