2,2-Bis[4-(4-maleimidophenoxy)phenyl]propane ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Bisphenol A bis(4-maleimidophenyl ether)
A334170-100G
A334170-25G
77550-950EA
91.67
USD
77550-950
77685-954
2,2-Bis[4-(4-maleimidophenoxy)phenyl]propane ≥98%
2,2-Bis[4-(4-maleimidophenoxy)phenyl]propane
Precaución: for Research Use Only
Formula:
C₃₅H₂₆N₂O₆ MW: 570.60 g/mol |
MDL Number:
MFCD09908251 CAS Number: 79922-55-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...