4-Methoxy-4'-trifluoromethylbenzophenone ≥97%

Supplier: Ambeed
Warning

A330041-250MG A330041-1G
77550-930EA 86 USD
77550-930 77593-950
4-Methoxy-4'-trifluoromethylbenzophenone ≥97%
4-Methoxy-4'-trifluoromethylbenzophenone

Precaución: for Research Use Only

Formula: C₁₅H₁₁F₃O₂
MDL Number: MFCD13152565
CAS Number: 6185-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR