tert-Butyl 3-(piperidin-4-yloxy)azetidine-1-carboxylate hydrochloride ≥97%

Supplier: Ambeed
Warning

A328652-250MG A328652-100MG A328652-1G
77550-916EA 240 USD
77550-916 77616-448 77748-644
tert-Butyl 3-(piperidin-4-yloxy)azetidine-1-carboxylate hydrochloride ≥97%
tert-Butyl 3-(piperidin-4-yloxy)azetidine-1-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₂₅ClN₂O₃
MDL Number: MFCD31381142
CAS Number: 2126176-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR