2-(Dichloromethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%

Supplier: Ambeed
Danger

A327457-25G A327457-5G A327457-10G A327457-1G
77550-912EA 1099 USD
77550-912 77550-914 77568-662 77622-282
2-(Dichloromethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-(Dichloromethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: For research use only.

Formula: C₇H₁₃BCl₂O₂
MDL Number: MFCD27976342
CAS Number: 83622-41-7
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR