Iodomethyl pivalate ≥98% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilized with Copper chip

A327304-25G A327304-5G
77550-910EA 24.72 USD
77550-910 77568-660
Iodomethyl pivalate ≥98% stabilized
Iodomethyl pivalate

Precaución: For research use only.

Formula: C₆H₁₁IO₂
MW: 242.05 g/mol
MDL Number: MFCD02179270
CAS Number: 53064-79-2
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR