Acrylophenone ≥95% stabilized

Supplier: Ambeed
Warning

Synonyms: 1-Phenylprop-2-en-1-one, 3-Oxo-3-phenylpropene

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A321863-1G A321863-250MG A321863-100MG
77550-894EA 145.23 USD
77550-894 77550-896 77568-648
Acrylophenone ≥95% stabilized
Acrylophenone

Precaución: For research use only.

Formula: C₉H₈O
MW: 132.159 g/mol
MDL Number: MFCD00457275
CAS Number: 768-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR