tert-Butyl(3-iodopropoxy)dimethylsilane ≥98%

Supplier: Ambeed
Danger

A307438-1G A307438-5G A307438-250MG
77550-842EA 22 USD
77550-842 77550-844 77885-817
tert-Butyl(3-iodopropoxy)dimethylsilane ≥98%
tert-Butyl(3-iodopropoxy)dimethylsilane

Precaución: for Research Use Only

Formula: C₉H₂₁IOSi
MDL Number: MFCD10569859
CAS Number: 78878-05-4
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR