Ricinoleic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: Ricinolic acid, (R)-12-Hydroxy-cis-9-octadecenoic acid, 12-Hydroxyoleic acid, Ricinelaidic acid, (R,Z)-12-Hydroxyoctadec-9-enoic acid

A299692-100G A299692-500G
77550-812EA 7 USD
77550-812 77601-344
Ricinoleic acid ≥98%
Ricinoleic acid

Precaución: for Research Use Only

Formula: C₁₈H₃₄O₃
MDL Number: MFCD00084840
CAS Number: 141-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR