(±)-4-Phenyl-2-butanol ≥99%

Supplier: Ambeed
Warning

A296951-25G A296951-5G
77550-800EA 54.59 USD
77550-800 77550-802
(±)-4-Phenyl-2-butanol ≥99%
(±)-4-Phenyl-2-butanol

Precaución: for Research Use Only

Formula: (C₆H₅)CH₂CH₂CH(OH)CH₃
MW: 150.22 g/mol
MDL Number: MFCD00044349
CAS Number: 2344-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR