4-Bromo-5-chloro-2-fluorobenzonitrile ≥98%

Supplier: Ambeed
Danger

A295286-1G A295286-5G
77550-792EA 667.44 USD
77550-792 77550-794
4-Bromo-5-chloro-2-fluorobenzonitrile ≥98%
4-Bromo-5-chloro-2-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂BrClFN
MW: 234.452 g/mol
MDL Number: MFCD19686183
CAS Number: 1349716-15-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR