Benzophenone-4,4'-dicarboxylic acid ≥98%

Supplier: Ambeed
Warning

A293620-5G A293620-25G
77550-782EA 51 USD
77550-782 77877-952
Benzophenone-4,4'-dicarboxylic acid ≥98%
Benzophenone-4,4'-dicarboxylic acid
Formula: C₁₅H₁₀O₅
MDL Number: MFCD00016538
CAS Number: 964-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR