3-Bromo-1,1,1-trifluoroacetone ≥98% (by GC)

Supplier: Ambeed
Danger

A292562-25G A292562-5G A292562-10G
77550-778EA 39.14 USD
77550-778 77612-686 77683-877
3-Bromo-1,1,1-trifluoroacetone ≥98% (by GC)
3-Bromo-1,1,1-trifluoroacetone

Precaución: For research use only.

Formula: BrH₂CC(O)CF₃
MW: 190.95 g/mol
MDL Number: MFCD00039237
CAS Number: 431-35-6
UN: 2924
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR