Chlorambucil ≥98%

Supplier: Ambeed
Danger

Synonyms: 4-(4-(Bis(2-chloroethyl)amino)phenyl)butanoic acid, 4-[4-[Bis(2-chloroethyl)amino]phenyl]butyric Acid

A285061-1G A285061-100MG A285061-250MG A285061-25G
77550-754EA 76.22 USD
77550-754 77571-772 77601-250 77672-470
Chlorambucil ≥98%
Chlorambucil

Precaución: for Research Use Only

Formula: C₁₄H₁₉Cl₂NO₂
MW: 304.21 g/mol
MDL Number: MFCD00021783
CAS Number: 305-03-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR