2-Amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: 3,5-Diiodotyrosine, H-DL-Tyr(3,5-DiI)-OH

A283913-1G A283913-100MG
77550-740EA 142 USD
77550-740 77685-806
2-Amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid ≥95%
2-Amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₉H₉I₂NO₃
MDL Number: MFCD00063076
CAS Number: 66-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR