3-(4-Methoxybenzoyl)acrylic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Methoxyphenyl)-4-oxobut-2-enoic acid

A277809-1G A277809-250MG
77550-710EA 41.2 USD
77550-710 77593-712
3-(4-Methoxybenzoyl)acrylic acid ≥95%
3-(4-Methoxybenzoyl)acrylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₄
MW: 206.20 g/mol
MDL Number: MFCD00020486
CAS Number: 5711-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR