N,O-Bis(trimethylsilyl)trifluoroacetamide ≥95%

Supplier: Ambeed
Danger

Synonyms: Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate, Bis(Trimethylsilyl)Trifluoroacetamide, BSTFA

A263628-25G A263628-5G
77550-654EA 6 USD
77550-654 77672-358
N,O-Bis(trimethylsilyl)trifluoroacetamide ≥95%
N,O-Bis(trimethylsilyl)trifluoroacetamide

Precaución: for Research Use Only

Formula: C₈H₁₈F₃NOSi₂
MDL Number: MFCD00008269
CAS Number: 25561-30-2
UN: 2924
ADR: 3,8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR