Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate ≥97%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyloxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid Pinacol ester, Benzyl 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate, 1-Cbz-1,2,3,6-tetrahydropyridine-4-boronic acid Pinacol ester, 1-Benzyloxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, Benzyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate, 1-Carbobenzoxy-1,2,3,6-tetrahydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-[1-(Carbobenzoxy)-1,2,3,6-tetrahydro-4-pyridyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, N-Cbz-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester

A258969-100G A258969-250MG
77550-632EA 257.5 USD
77550-632 77672-306
Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate ≥97%
Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₉H₂₆BNO₄
MW: 343.23 g/mol
MDL Number: MFCD11521562
CAS Number: 286961-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR