2-(3-Bromo-5-chlorophenyl)acetonitrile ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A253355-5G A253355-1G
77550-610EA 328.57 USD
77550-610 77683-761
2-(3-Bromo-5-chlorophenyl)acetonitrile ≥98%, Ambeed.Inc
2-(3-Bromo-5-chlorophenyl)acetonitrile

Precaución: For research use only.

Formula: C₈H₅BrClN
MW: 230.489 g/mol
MDL Number: MFCD13195464
CAS Number: 1056454-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR