Tris(2,2,2-trifluoroethyl) phosphite 98%

Supplier: Ambeed
Warning

A251708-5G A251708-1G
77550-596EA 45 USD
77550-596 77568-444
Tris(2,2,2-trifluoroethyl) phosphite 98%
Tris(2,2,2-trifluoroethyl) phosphite

Precaución: For research use only.

Formula: C₆H₆F₉O₃P
MDL Number: MFCD00009902
CAS Number: 370-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR