tert-Butyl 4-(piperidin-4-ylmethyl)piperidine-1-carboxylate ≥98%

Supplier: Ambeed
Warning

Synonyms: tert-butyl 4-[(piperidin-4-yl)methyl]piperidine-1-carboxylate

A240840-1G A240840-250MG A240840-100MG
77550-552EA 205 USD
77550-552 77612-338 77892-427
tert-Butyl 4-(piperidin-4-ylmethyl)piperidine-1-carboxylate ≥98%
tert-Butyl 4-(piperidin-4-ylmethyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₆H₃₀N₂O₂
MDL Number: MFCD15474945
CAS Number: 879883-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR