[2,2'-Bipyridine]-6-carboximidamide hydrochloride ≥95%

Supplier: Ambeed
Warning

A240660-1G A240660-250MG
77550-546EA 296.64 USD
77550-546 77593-538
[2,2'-Bipyridine]-6-carboximidamide hydrochloride ≥95%
[2,2'-Bipyridine]-6-carboximidamide hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClN₄
MW: 234.68 g/mol
MDL Number: MFCD12755605
CAS Number: 219745-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR