n-Dodecyl acrylate ≥98% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A239257-25G A239257-100G
77550-542EA 5 USD
77550-542 77879-417
n-Dodecyl acrylate ≥98% stabilized
n-Dodecyl acrylate

Precaución: for Research Use Only

Formula: C₁₅H₂₈O₂
MDL Number: MFCD00042871
CAS Number: 2156-97-0
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR