3-(3-Methyl-3H-diazirin-3-yl)propanoic acid ≥97%

Supplier: Ambeed
Danger

A236989-100MG A236989-1G A236989-250MG
77550-524EA 104.03 USD
77550-524 77550-526 77550-528
3-(3-Methyl-3H-diazirin-3-yl)propanoic acid ≥97%
3-(3-Methyl-3H-diazirin-3-yl)propanoic acid

Precaución: for Research Use Only

MDL Number: MFCD19228230
CAS Number: 25055-86-1
UN: 2924
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR