Poly(allylamine hydrochloride), M.W. ~15000 ≥97%

Supplier: Ambeed
Warning

A227086-5G A227086-100G
77550-468EA 7.21 USD
77550-468 77568-198
Poly(allylamine hydrochloride), M.W. ~15000 ≥97%
Poly(allylamine hydrochloride)

Precaución: For research use only.

Formula: [CH₂CH(CH₂NH₂·HCl)]n
MW: 93.5600 g/mol
MDL Number: MFCD00084396
CAS Number: 71550-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR