Methyl 4-oxobutanoate ≥95%

Supplier: Ambeed
Warning

A224007-1G A224007-5G A224007-100MG A224007-250MG
77550-452EA 56.65 USD
77550-452 77550-454 77593-458 77615-862
Methyl 4-oxobutanoate ≥95%
Methyl 4-oxobutanoate

Precaución: for Research Use Only

Formula: C₅H₈O₃
MW: 116.11 g/mol
MDL Number: MFCD00082185
CAS Number: 13865-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR