Mono-(6-ethanediamine-6-deoxy)-β-Cyclodextrin ≥98%

Supplier: Ambeed
Warning

A220626-1G A220626-5G
77550-430EA 115 USD
77550-430 77901-757
Mono-(6-ethanediamine-6-deoxy)-β-Cyclodextrin ≥98%
Mono-(6-ethanediamine-6-deoxy)-β-Cyclodextrin

Precaución: for Research Use Only

Formula: C₄₄H₇₆N₂O₃₄
MDL Number: MFCD22200979
CAS Number: 60984-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR