Amberlyst® 15 ≤1.5% in water, dry ion exchange resin ≤1.5%H2O

Supplier: Ambeed
Warning

A218148-25G A218148-100G A218148-1000G A218148-500G
77550-412EA 14 USD
77550-412 77568-116 77877-681 77877-682
Amberlyst® 15 ≤1.5% in water, dry ion exchange resin ≤1.5%H2O
Amberlyst® 15

Precaución: For research use only.

MDL Number: MFCD00145841
CAS Number: 9037-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR