2-Chloro-3,4-dihydroxybenzoic acid 95%

Supplier: Ambeed
Warning

A215020-1G A215020-5G A215020-250MG
77550-390EA 115.36 USD
77550-390 77568-076 77683-614
2-Chloro-3,4-dihydroxybenzoic acid 95%
2-Chloro-3,4-dihydroxybenzoic acid

Precaución: For research use only.

Formula: C₇H₅ClO₄
MW: 188.56 g/mol
MDL Number: MFCD20640292
CAS Number: 87932-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR