(S)-(+)-2,2-Dimethylcyclopropanecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2,2-Dimethylcyclopropanecarboxylic acid

A208996-10G A208996-1G A208996-5G A208996-25G
77550-348EA 346.08 USD
77550-348 77568-008 77568-010 77685-540
(S)-(+)-2,2-Dimethylcyclopropanecarboxylic acid 97%
(S)-(+)-2,2-Dimethylcyclopropanecarboxylic acid

Precaución: For research use only.

Formula: C₆H₁₀O₂
MW: 114.1400 g/mol
MDL Number: MFCD08460918
CAS Number: 14590-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR