Dichloro[1,3-bis(2,6-Di-3-pentylphenyl)imidazol-2-ylidene](3-chloropyridyl)palladium(II) 98%

Supplier: Ambeed
Warning

A207902-100MG A207902-1G A207902-250MG A207902-5G A207902-50MG
77550-336EA 83 USD
77550-336 77550-338 77550-340 77567-994 77746-277
Dichloro[1,3-bis(2,6-Di-3-pentylphenyl)imidazol-2-ylidene](3-chloropyridyl)palladium(II) 98%
Dichloro[1,3-bis(2,6-Di-3-pentylphenyl)imidazol-2-ylidene](3-chloropyridyl)palladium(II)

Precaución: For research use only.

Formula: C₄₀H₅₆Cl₃N₃Pd
MDL Number: MFCD18253025
CAS Number: 1158652-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR